Gene Transcripts

Gene Symbol
Myo18b
Entrez Gene ID
74376
Gene Name
myosin XVIIIb
Gene Aliases
4932408L24Rik, 4933411E19Rik, Gm448
Chromosome Location
Chr. 5: 112688876 - 112896459 on Build GRCm38
UniGene ID
Mm.441903
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028901.2 NP_083177.2 26, 27 4781
GenBank mRNA BC145218.1 - 20, 21 2730

View Full Product Details