Gene Transcripts

Gene Symbol
Snx27
Entrez Gene ID
76742
Gene Name
sorting nexin family member 27
Gene Aliases
5730552M22Rik, ESTM45, ESTM47, R75405
Chromosome Location
Chr. 3: 94497542 - 94582716 on Build GRCm38
UniGene ID
Mm.28160
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082484.2 NP_001075953.1 2 620
NM_029721.2 NP_083997.1 2 620
GenBank mRNA AK122296.1 - 2 432
AK147452.1 - 2 607
BC053495.1 - 2 432

View Full Product Details