Gene Transcripts

Gene Symbol
Krtap5-3
Entrez Gene ID
77215
Gene Name
keratin associated protein 5-3
Gene Aliases
A030007E19Rik
Chromosome Location
Chr. 7: 142201364 - 142203015 on Build GRCm38
UniGene ID
Mm.462993
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023860.1 NP_076349.1 1 1349
GenBank mRNA AK020699.1 - 2 772

View Full Product Details