Gene Transcripts
- Gene Symbol
- Notch2
- Entrez Gene ID
- 29492
- Gene Name
- notch 2
- Gene Aliases
- Chromosome Location
- Chr. 2: 188198299 - 188200386 on Build Rnor_6.0
- UniGene ID
- Rn.65930
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024358.1 |
NP_077334.1
|
3, 4 |
933 |
|
GenBank mRNA |
M93661.1 |
- |
3, 4 |
933 |
|
View Full Product Details