Gene Transcripts

Gene Symbol
Suox
Entrez Gene ID
81805
Gene Name
sulfite oxidase
Gene Aliases
Chromosome Location
Chr. 7: 3098228 - 3102168 on Build Rnor_6.0
UniGene ID
Rn.25720
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031127.3 NP_112389.3 3 388
XM_006240802.2 XP_006240864.1 4 468
XM_006240803.2 XP_006240865.1 5 569
GenBank mRNA BC061991.1 - 3 388
L05084.1 - 2 244

View Full Product Details