Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145097.2 | NP_659565.2 | 2 | 345 | |
XM_008764741.1 | XP_008762963.1 | 3 | 573 | ||
GenBank mRNA | BC088111.1 | - | 2 | 345 | |
FQ209558.1 | - | 2 | 362 | ||
FQ210636.1 | - | 2 | 365 | ||
FQ218781.1 | - | 1 | 113 | ||
FQ218835.1 | - | 2 | 365 | ||
FQ219263.1 | - | 1 | 113 | ||
FQ219393.1 | - | 2 | 415 | ||
FQ219554.1 | - | 1 | 113 | ||
U51017.1 | - | 1 | 306 |