Gene Transcripts

Gene Symbol
Slc34a1
Entrez Gene ID
25548
Gene Name
solute carrier family 34 (type II sodium/phosphate cotransporter), member 1
Gene Aliases
NaPi-2, NaPi2A, Npt2, Slc17a2
Chromosome Location
Chr. 17: 9747766 - 9762739 on Build Rnor_6.0
UniGene ID
Rn.92022
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013030.1 NP_037162.1 8, 9 986
GenBank mRNA BC078876.1 - 8, 9 980
L13257.1 - 8, 9 986

View Full Product Details