Gene Transcripts

Gene Symbol
Npff
Entrez Gene ID
60337
Gene Name
neuropeptide FF-amide peptide precursor
Gene Aliases
Chromosome Location
Chr. 7: 144127084 - 144127873 on Build Rnor_6.0
UniGene ID
Rn.48705
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022586.1 NP_072108.1 2 222
GenBank mRNA AF148700.1 - 2 222

View Full Product Details