Gene Transcripts

Gene Symbol
Frat1
Entrez Gene ID
14296
Gene Name
frequently rearranged in advanced T cell lymphomas
Gene Aliases
AW060382
Chromosome Location
Chr. 19: 41829970 - 41832583 on Build GRCm38
UniGene ID
Mm.4573
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008043.3 NP_032069.2 1 1224
GenBank mRNA AK015680.1 - 1 1158
BC140987.1 - 1 1078 9055599
U58974.2 - 1 1077

View Full Product Details