Gene Transcripts

Gene Symbol
CSRNP1
Entrez Gene ID
64651
Gene Name
cysteine and serine rich nuclear protein 1
Gene Aliases
AXUD1, CSRNP-1, FAM130B, TAIP-3, URAX1
Chromosome Location
Chr. 3: 39141851 - 39154602 on Build GRCh38
UniGene ID
Hs.370950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320559.1 NP_001307488.1 3 791
NM_001320560.1 NP_001307489.1 3 592
NM_033027.3 NP_149016.2 3 660
XM_017007048.1 XP_016862537.1 3 3756
GenBank mRNA AB053121.1 - 3 637
AB063301.1 - 3 552
BC038949.1 - 3 607 5547938
BG536908.1 - 3 659
BX397882.2 - 3 592

View Full Product Details