Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253804.1 | NP_001240733.1 | 2 | 210 | |
NM_009195.3 | NP_033221.1 | 2 | 204 | ||
GenBank mRNA | AF047339.1 | - | 2 | 128 | |
AF121118.1 | - | 2 | 195 | ||
AK149969.1 | - | 2 | 193 | ||
AK151717.1 | - | 2 | 165 | ||
AK159274.1 | - | 2 | 192 | ||
AK159331.1 | - | 2 | 187 | ||
AK159483.1 | - | 2 | 160 | ||
AK159537.1 | - | 2 | 187 | ||
AK160034.1 | - | 2 | 153 | ||
AK160072.1 | - | 2 | 187 | ||
AK164479.1 | - | 2 | 169 | ||
AK165874.1 | - | 2 | 182 | ||
AK166304.1 | - | 2 | 163 | ||
AK167371.1 | - | 2 | 201 | ||
AK167444.1 | - | 2 | 201 | ||
AK169044.1 | - | 2 | 159 | ||
BC066872.1 | - | 2 | 134 | 30436525 | |
BY740705.1 | - | 2 | 205 |