Gene Transcripts

Gene Symbol
Sar1a
Entrez Gene ID
20224
Gene Name
SAR1 gene homolog A (S. cerevisiae)
Gene Aliases
1600019H17Rik, Sara, Sara1
Chromosome Location
Chr. 10: 61680304 - 61693297 on Build GRCm38
UniGene ID
Mm.391794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009120.2 NP_033146.1 7 797
GenBank mRNA AK157843.1 - 7 809
AK159239.1 - 7 808
AK194825.1 - 1 118
AK206663.1 - 1 118
BC005549.1 - 7 798 3585667


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK129305.1 - 3995 Chr. - - [-]

View Full Product Details