Gene Transcripts

Gene Symbol
Smarcc1
Entrez Gene ID
20588
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 1
Gene Aliases
AI115498, BAF155, Rsc8, SRG3
Chromosome Location
Chr. 9: 110132024 - 110240178 on Build GRCm38
UniGene ID
Mm.85410
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009211.2 NP_033237.2 28 3900
GenBank mRNA AK143485.1 - 28 3888
AK144122.1 - 29 3987
U85614.1 - 28 3853

View Full Product Details