Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001271603.1 | NP_001258532.1 | 2 | 1241 | |
NM_009896.2 | NP_034026.1 | 2 | 1121 | ||
GenBank mRNA | AB000677.1 | - | 1 | 965 | |
AF120490.1 | - | 2 | 1094 | ||
AF180302.1 | - | 2 | 1099 | ||
AK028632.1 | - | 1122 | |||
AK154706.1 | - | 1138 | |||
U88325.1 | - | 2 | 1083 |