Gene Transcripts

Gene Symbol
Prl7a2
Entrez Gene ID
19114
Gene Name
prolactin family 7, subfamily a, member 2
Gene Aliases
PLP-F, Prlpf
Chromosome Location
Chr. 13: 27658584 - 27668052 on Build GRCm38
UniGene ID
Mm.153999
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011168.4 NP_035298.1 6 609
GenBank mRNA AF011382.1 - 6 598
AF020524.1 - 6 605
AK005455.1 - 6 610
AK028068.1 - 6 609
AK131665.1 - 6 610
BC099468.1 - 6 588 30789902

View Full Product Details