Gene Transcripts

Gene Symbol
YES1
Entrez Gene ID
7525
Gene Name
YES proto-oncogene 1, Src family tyrosine kinase
Gene Aliases
HsT441, P61-YES, Yes, c-yes
Chromosome Location
Chr. 18: 721592 - 812326 on Build GRCh38
UniGene ID
Hs.194148
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005433.3 NP_005424.1 2 373
XM_005258139.3 XP_005258196.1 2 335
XM_017025960.1 XP_016881449.1 2 215
GenBank mRNA AK314063.1 - 2 317
BC048960.1 - 2 369 5260751
M15990.1 - 2 359

View Full Product Details