Gene Transcripts
- Gene Symbol
- Rpl9
- Entrez Gene ID
- 20005
- Gene Name
- ribosomal protein L9
- Gene Aliases
- Chromosome Location
- Chr. 5: 65388364 - 65391431 on Build GRCm38
- UniGene ID
- Mm.300271
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011292.2 |
NP_035422.1
|
3, 4 |
226 |
|
GenBank mRNA |
AF260271.1 |
- |
2, 3 |
196 |
|
AK012331.1 |
- |
3, 4 |
196 |
|
AK012445.1 |
- |
3, 4 |
196 |
|
AK017388.1 |
- |
3, 4 |
196 |
|
AK017427.1 |
- |
3, 4 |
196 |
|
AK053431.1 |
- |
2, 3 |
579 |
|
AK084278.1 |
- |
3, 4 |
197 |
|
AK088166.1 |
- |
3, 4 |
196 |
|
AK153562.1 |
- |
3, 4 |
196 |
|
AK159980.1 |
- |
3, 4 |
196 |
|
AK168326.1 |
- |
3, 4 |
194 |
|
AK169217.1 |
- |
3, 4 |
194 |
|
BC013165.1 |
- |
2, 3 |
186 |
2655358 |
BC081435.1 |
- |
2, 3 |
172 |
6824107 |
BC083166.1 |
- |
2, 3 |
177 |
6397429 |
BC083329.1 |
- |
2, 3 |
175 |
6824346 |
BC086937.1 |
- |
3, 4 |
193 |
6787671 |
BC089319.1 |
- |
2, 3 |
175 |
6823996 |
BC093526.1 |
- |
2, 3 |
175 |
5150493 |
U17331.1 |
- |
1, 2 |
132 |
|
U17332.1 |
- |
1, 2 |
132 |
|
View Full Product Details