Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253904.1 | NP_001240833.1 | 48 | 8386 | |
NM_001253905.1 | NP_001240834.1 | 43 | 7835 | ||
NM_011424.3 | NP_035554.3 | 47 | 8434 | ||
GenBank mRNA | AF125671.1 | - | 47 | 8242 | |
AK147308.1 | - | 27 | 5361 | ||
AK147394.1 | - | 48 | 8351 | ||
AK161209.1 | - | 1 | 736 | ||
AK168836.1 | - | 1 | 765 | ||
BC047524.1 | - | 11 | 2599 |