Gene Transcripts

Gene Symbol
Gpc6
Entrez Gene ID
23888
Gene Name
glypican 6
Gene Aliases
Chromosome Location
Chr. 14: 116921001 - 117979529 on Build GRCm38
UniGene ID
Mm.440025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079844.2 NP_001073313.1 10 2783
NM_011821.3 NP_035951.1 9 2753
GenBank mRNA AF105268.1 - 10 2480
AK164668.1 - 9 2755

View Full Product Details