Gene Transcripts

Gene Symbol
Mid2
Entrez Gene ID
23947
Gene Name
midline 2
Gene Aliases
FXY2, Trim1
Chromosome Location
Chr. X: 140664303 - 140767715 on Build GRCm38
UniGene ID
Mm.131097
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011845.2 NP_035975.1 3, 4 896
GenBank mRNA AF196480.1 - 3, 4 896
Y18881.1 - 4, 5 1251

View Full Product Details