Gene Transcripts
- Gene Symbol
- Hnrnpc
- Entrez Gene ID
- 15381
- Gene Name
- heterogeneous nuclear ribonucleoprotein C
- Gene Aliases
- AL022939, D14Wsu171e, Hnrpc, Hnrpc1, Hnrpc2, hnRNPC1, hnRNPC2, hnrnp-C
- Chromosome Location
- Chr. 14: 52073380 - 52104028 on Build GRCm38
- UniGene ID
- Mm.427321
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001170981.1 |
NP_001164452.1
|
3, 4 |
444 |
|
NM_001170982.1 |
NP_001164453.1
|
2, 3 |
418 |
|
NM_001170983.1 |
NP_001164454.1
|
3, 4 |
444 |
|
NM_001170984.1 |
NP_001164455.1
|
3, 4 |
444 |
|
NM_016884.3 |
NP_058580.1
|
3, 4 |
444 |
|
GenBank mRNA |
AF095257.1 |
- |
3, 4 |
398 |
|
AK010845.1 |
- |
3, 4 |
423 |
|
AK011336.1 |
- |
3, 4 |
416 |
|
AK012633.2 |
- |
3, 4 |
406 |
|
AK019958.1 |
- |
3, 4 |
1619 |
|
AK088245.1 |
- |
3, 4 |
421 |
|
AK088678.1 |
- |
3, 4 |
416 |
|
AK089061.1 |
- |
3, 4 |
424 |
|
AK151446.1 |
- |
3, 4 |
421 |
|
AK153053.1 |
- |
3, 4 |
421 |
|
AK166590.1 |
- |
3, 4 |
422 |
|
AK168802.1 |
- |
3, 4 |
418 |
|
AK169287.1 |
- |
3, 4 |
416 |
|
AK184811.1 |
- |
3, 4 |
351 |
|
AK212671.1 |
- |
1, 2 |
270 |
|
AK214960.1 |
- |
2, 3 |
306 |
|
BC004706.1 |
- |
3, 4 |
406 |
3499283 |
BC095922.1 |
- |
2, 3 |
359 |
4988965 |
CK793036.1 |
- |
|
398 |
|
DV652725.1 |
- |
3, 4 |
468 |
|
View Full Product Details