Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001110310.1 | NP_001103780.1 | 3 | 470 | |
NM_001110311.1 | NP_001103781.1 | 3 | 470 | ||
NM_018875.2 | NP_061363.2 | 3 | 470 | ||
GenBank mRNA | AF062484.1 | - | 3 | 394 | |
AK011257.1 | - | 3 | 361 | ||
AK049561.1 | - | 3 | 363 | ||
AK131838.1 | - | 3 | 460 | ||
AK146833.1 | - | 3 | 390 | ||
AK148235.1 | - | 3 | 423 | ||
AK168604.1 | - | 3 | 361 | ||
AK169569.1 | - | 3 | 446 | ||
AK170403.1 | - | 3 | 361 | ||
AK170992.1 | - | 3 | 457 | ||
BC002009.1 | - | 3 | 399 | 3592627 |