Gene Transcripts
- Gene Symbol
- Tyro3
- Entrez Gene ID
- 22174
- Gene Name
- TYRO3 protein tyrosine kinase 3
- Gene Aliases
- AI323366, Brt, Dtk, Etk-2, Rse, Sky, TK19-2, Tif, etk2/tyro3
- Chromosome Location
- Chr. 2: 119797740 - 119818104 on Build GRCm38
- UniGene ID
- Mm.2901
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290800.1 |
NP_001277729.1
|
18, 19 |
2995 |
|
NM_019392.2 |
NP_062265.2
|
18, 19 |
2531 |
|
GenBank mRNA |
AB000828.1 |
- |
18, 19 |
2268 |
|
AK134607.1 |
- |
7, 8 |
2079 |
|
AK141198.1 |
- |
18, 19 |
2302 |
|
AK213086.1 |
- |
2, 3 |
240 |
|
BC066058.1 |
- |
18, 19 |
2531 |
6834015 |
BC082325.1 |
- |
18, 19 |
2479 |
30356080 |
D17393.1 |
- |
18, 19 |
2973 |
|
U05683.1 |
- |
18, 19 |
2316 |
|
U18342.1 |
- |
18, 19 |
2448 |
|
U18343.1 |
- |
18, 19 |
2991 |
|
U18933.1 |
- |
18, 19 |
2467 |
|
X78103.1 |
- |
18, 19 |
2475 |
|
View Full Product Details