Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019586.3 | NP_062532.2 | 2, 3 | 274 | |
GenBank mRNA | AJ245899.1 | - | 2, 3 | 208 | |
AK003050.1 | - | 2, 3 | 206 | ||
AK031669.1 | - | 2, 3 | 162 | ||
AK149985.1 | - | 2, 3 | 185 | ||
AK160428.1 | - | 2, 3 | 280 | ||
BC024623.1 | - | 2, 3 | 212 | 4217620 | |
BY707310.1 | - | 2, 3 | 274 |