Gene Transcripts

Gene Symbol
PKD2L2
Entrez Gene ID
27039
Gene Name
polycystin 2 like 2, transient receptor potential cation channel
Gene Aliases
TRPP5
Chromosome Location
Chr. 5: 137889420 - 137942747 on Build GRCh38
UniGene ID
Hs.716884
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258448.1 NP_001245377.1 4 356
NM_001258449.1 NP_001245378.1 4 356
NM_001300921.1 NP_001287850.1 4 356
NM_014386.3 NP_055201.2 4 356
XM_011543318.2 XP_011541620.1 3 270
XM_011543321.1 XP_011541623.1 4 290
XM_017009343.1 XP_016864832.1 4 372
XM_017009344.1 XP_016864833.1 3 480
XM_017009345.1 XP_016864834.1 4 372
GenBank mRNA AF118125.1 - 4 323
AF182034.1 - 4 326
AK301924.1 - 4 331
BC034619.1 - 4 1409
BC038714.1 - 4 292
BC044581.1 - 4 355 4830805
BG719682.1 - 4 320
BG721989.1 - 4 361

View Full Product Details