Gene Transcripts

Gene Symbol
Prl8a9
Entrez Gene ID
67310
Gene Name
prolactin family8, subfamily a, member 9
Gene Aliases
1600019F10Rik, PLP-Cbeta, Prlpc2
Chromosome Location
Chr. 13: 27558001 - 27564616 on Build GRCm38
UniGene ID
Mm.46091
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023332.3 NP_075821.1 6 672
GenBank mRNA AF158744.1 - 6 657
AK005409.1 - 6 673
AK005494.1 - 6 673
BC064762.1 - 6 640 30013931

View Full Product Details