Gene Transcripts
- Gene Symbol
- U2af1
- Entrez Gene ID
- 108121
- Gene Name
- U2 small nuclear ribonucleoprotein auxiliary factor (U2AF) 1
- Gene Aliases
- 2010107D16Rik, 35kDa
- Chromosome Location
- Chr. 17: 31647081 - 31658764 on Build GRCm38
- UniGene ID
- Mm.379289
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163769.1 |
NP_001157241.1
|
4, 5 |
315 |
|
NM_024187.4 |
NP_077149.2
|
4, 5 |
315 |
|
GenBank mRNA |
AK008332.1 |
- |
4, 5 |
277 |
|
AK012334.1 |
- |
4, 5 |
275 |
|
AK012849.1 |
- |
4, 5 |
275 |
|
AK088979.1 |
- |
4, 5 |
285 |
|
AK150872.1 |
- |
4, 5 |
315 |
|
AV453783.1 |
- |
4, 5 |
297 |
|
BC002184.1 |
- |
4, 5 |
254 |
|
BC030413.1 |
- |
1, 2 |
1116 |
|
BC033478.1 |
- |
2, 3 |
83 |
|
BC069888.1 |
- |
4, 5 |
257 |
|
BC115479.1 |
- |
4, 5 |
277 |
40100558 |
BC115480.1 |
- |
4, 5 |
277 |
40100559 |
View Full Product Details