Gene Transcripts

Gene Symbol
Mrpl16
Entrez Gene ID
94063
Gene Name
mitochondrial ribosomal protein L16
Gene Aliases
2310039D06Rik
Chromosome Location
Chr. 19: 11770415 - 11774956 on Build GRCm38
UniGene ID
Mm.203928
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025606.3 NP_079882.2 4 1023
GenBank mRNA AB049643.1 - 4 1023
AK009687.1 - 4 1008
BC044721.1 - 4 1051 6476049

View Full Product Details