Gene Transcripts
- Gene Symbol
- Ubr7
- Entrez Gene ID
- 66622
- Gene Name
- ubiquitin protein ligase E3 component n-recognin 7 (putative)
- Gene Aliases
- 5730410I19Rik, AA589405, AW557761
- Chromosome Location
- Chr. 12: 102757975 - 102777701 on Build GRCm38
- UniGene ID
- Mm.34261
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025666.2 |
NP_079942.1
|
4, 5 |
503 |
|
GenBank mRNA |
AK011968.1 |
- |
4, 5 |
374 |
|
AK017542.1 |
- |
4, 5 |
459 |
|
AK088208.1 |
- |
4, 5 |
505 |
|
AK146308.1 |
- |
4, 5 |
504 |
|
AK149989.1 |
- |
4, 5 |
458 |
|
AK155611.1 |
- |
4, 5 |
374 |
|
AK190790.1 |
- |
3, 4 |
237 |
|
BC051678.1 |
- |
1, 2 |
31 |
|
BC058535.1 |
- |
3, 4 |
187 |
|
BC079531.1 |
- |
3, 4 |
200 |
|
BC094323.1 |
- |
4, 5 |
510 |
|
BC094423.1 |
- |
4, 5 |
461 |
6812389 |
View Full Product Details