Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025812.2 | NP_080088.1 | 3 | 287 | |
GenBank mRNA | AK004596.1 | - | 3 | 307 | |
AK017716.1 | - | 3 | 490 | ||
AK032790.1 | - | 3 | 315 | ||
AK039222.1 | - | 3 | 309 | ||
AK086346.1 | - | 3 | 305 | ||
AK129010.1 | - | 3 | 489 | ||
AK134213.1 | - | 3 | 459 | ||
AK154075.1 | - | 3 | 306 | ||
AL355705.1 | - | 3 | 273 | ||
BC013804.1 | - | 3 | 287 | 3602239 | |
BC068257.1 | - | 3 | 360 | 6434284 | |
DQ182735.1 | - | 3 | 287 |