Gene Transcripts

Gene Symbol
AGXT
Entrez Gene ID
189
Gene Name
alanine-glyoxylate aminotransferase
Gene Aliases
AGT, AGT1, AGXT1, PH1, SPAT, SPT, TLH6
Chromosome Location
Chr. 2: 240868745 - 240879119 on Build GRCh38
UniGene ID
Hs.144567
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000030.2 NP_000021.1 6, 7 804
GenBank mRNA AB292648.1 - 6, 7 705
AF348451.1 - 6, 7 595
AK292754.1 - 6, 7 725
AK315331.1 - 6, 7 752
BC132819.1 - 6, 7 801 40146841
D13368.1 - 6, 7 791
X53414.1 - 6, 7 805
X56092.1 - 6, 7 704

View Full Product Details