Gene Transcripts
- Gene Symbol
- Wls
- Entrez Gene ID
- 68151
- Gene Name
- wntless homolog (Drosophila)
- Gene Aliases
- 5031439A09Rik, AI173978, AI987742, Gpr177
- Chromosome Location
- Chr. 3: 159839677 - 159938670 on Build GRCm38
- UniGene ID
- Mm.6766
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026582.4 |
NP_080858.3
|
4, 5 |
906 |
|
GenBank mRNA |
AK019886.1 |
- |
5, 6 |
945 |
|
AK029053.1 |
- |
4, 5 |
865 |
|
AK029104.1 |
- |
4, 5 |
863 |
|
AK029315.1 |
- |
4, 5 |
863 |
|
AK031444.1 |
- |
4, 5 |
863 |
|
AK044810.1 |
- |
4, 5 |
865 |
|
AK049842.1 |
- |
4, 5 |
863 |
|
AK077407.1 |
- |
4, 5 |
864 |
|
AK128912.1 |
- |
4, 5 |
862 |
|
AK159207.1 |
- |
4, 5 |
863 |
|
AK159977.1 |
- |
4, 5 |
865 |
|
AK190589.1 |
- |
1, 2 |
62 |
|
BC018381.1 |
- |
3, 4 |
490 |
4237619 |
BC075615.1 |
- |
4, 5 |
906 |
5703610 |
View Full Product Details