Gene Transcripts

Gene Symbol
Slc35a5
Entrez Gene ID
74102
Gene Name
solute carrier family 35, member A5
Gene Aliases
1010001J06Rik, AU021179, BB097433, D16Ertd450e, D730043G07Rik
Chromosome Location
Chr. 16: 45139573 - 45158673 on Build GRCm38
UniGene ID
Mm.264984
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028756.4 NP_083032.2 7 3231
GenBank mRNA AK033921.1 - 7 3200

View Full Product Details