Gene Transcripts

Gene Symbol
Mtmr3
Entrez Gene ID
74302
Gene Name
myotubularin related protein 3
Gene Aliases
1700092A20Rik, AI255150, AW557713, FYVE-DSP1, ZFYVE10, mKIAA0371
Chromosome Location
Chr. 11: 4480868 - 4594815 on Build GRCm38
UniGene ID
Mm.425669
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028860.2 NP_083136.2 7 781
GenBank mRNA AK141138.1 - 7 761
BC032166.1 - 7 764 5352919
BC081544.1 - 7 765 3672099

View Full Product Details