Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033074.3 | NP_149065.2 | 2 | 212 | |
GenBank mRNA | AK010256.1 | - | 2 | 179 | |
AK011146.2 | - | 2 | 213 | ||
AK031064.1 | - | 2 | 176 | ||
AK152945.1 | - | 2 | 173 | ||
AK153314.1 | - | 2 | 184 | ||
AK166693.1 | - | 2 | 179 | ||
AK167597.1 | - | 2 | 221 | ||
AK168969.1 | - | 2 | 172 | ||
AK206659.1 | - | 1 | 18 | ||
AK210237.1 | - | 1 | 18 | ||
BC055371.1 | - | 2 | 198 | 30060749 | |
BC060176.1 | - | 155 |