Gene Transcripts

Gene Symbol
Tmem2
Entrez Gene ID
83921
Gene Name
transmembrane protein 2
Gene Aliases
Chromosome Location
Chr. 19: 21777508 - 21858437 on Build GRCm38
UniGene ID
Mm.329776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033759.2 NP_001028931.1 13 2841
NM_031997.4 NP_114386.3 14 2923
GenBank mRNA AK129352.1 - 8 1142
BC019745.1 - 5 564 3675225
BC076570.1 - 13 2447 6830791
BC089353.1 - 13 2823 6410437
BU058172.1 - 4 355
CA752446.1 - 6 781

View Full Product Details