Gene Transcripts

Gene Symbol
Rtn3
Entrez Gene ID
20168
Gene Name
reticulon 3
Gene Aliases
RTN3-A1
Chromosome Location
Chr. 19: 7425895 - 7483291 on Build GRCm38
UniGene ID
Mm.246990
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003933.2 NP_001003933.1 3, 4 2694
NM_001003934.2 NP_001003934.1 4, 5 2751
NM_001271486.1 NP_001258415.1 4, 5 1788
NM_001271487.1 NP_001258416.1 3, 4 627
NM_053076.3 NP_444306.1 2, 3 570
GenBank mRNA AB046114.1 - 2, 3 564
AF195940.1 - 2, 3 514
AK032109.1 - 4, 5 1765
AK036892.1 - 2, 3 536
AK049845.1 - 2, 3 502
AK075883.1 - 2, 3 499
AK088670.1 - 2, 3 502
AK146505.1 - 2, 3 378
AK148792.1 - 2, 3 529
AK148947.1 - 2, 3 531
AK165683.1 - 2, 3 501
AK197640.1 - 1, 2 220
AK217412.1 - 1, 2 127
AK218971.1 - 1, 2 136
AY164700.1 - 2, 3 564
AY427822.1 - 3, 4 2675
AY507126.1 - 2, 3 362
AY507127.1 - 2, 3 2297
AY750849.1 - 4, 5 2732
BC005478.1 - 2, 3 473
BC014697.1 - 2, 3 481
BC036717.1 - 2, 3 479
BC079882.1 - 4, 5 2661 6839323
BI729783.1 - 541

View Full Product Details