Gene Transcripts

Gene Symbol
PON1
Entrez Gene ID
5444
Gene Name
paraoxonase 1
Gene Aliases
ESA, MVCD5, PON
Chromosome Location
Chr. 7: 95298357 - 95324572 on Build GRCh38
UniGene ID
Hs.370995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA D84371.1 - 9 2197
U53784.1 - 9 2197
Z70723.1 - 9 2197

View Full Product Details