Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133791.4 | NP_598552.2 | 11 | 2217 | |
GenBank mRNA | AB045323.1 | - | 11 | 2242 | |
AK032989.1 | - | 7 | 1283 | ||
AK079375.1 | - | 11 | 2053 | ||
AK086956.1 | - | 11 | 2047 | ||
AK147648.1 | - | 2211 | |||
AK199576.1 | - | 1 | 79 | ||
BC054434.1 | - | 1 | 498 | ||
BC067050.1 | - | 11 | 2217 | 30537299 |