Gene Transcripts

Gene Symbol
RPL9
Entrez Gene ID
6133
Gene Name
ribosomal protein L9
Gene Aliases
L9, NPC-A-16
Chromosome Location
Chr. 4: 39454124 - 39458948 on Build GRCh38
UniGene ID
Hs.412370
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000661.4 NP_000652.2 4, 5 313
NM_001024921.3 NP_001020092.1 4, 5 471
GenBank mRNA AB062431.1 - 1 260
AK130966.1 - 1 276
AK297178.1 - 4, 5 296
AK303908.1 - 1 276
AK312241.1 - 1 280
AL039355.2 - 4, 5 446
AY320412.1 - 4, 5 296
AY376242.1 - 3, 4 430
AY597415.1 - 1 287
BC000483.2 - 1 265 2964733
BC004156.2 - 1 259 2961026
BC004206.2 - 1 259 2961026
BC007967.2 - 1 315 4304670
BC012149.1 - 1 315 4554838
BC031906.1 - 3, 4 260 4996986
BC066318.1 - 1 280 5277491
BC070214.1 - 1 319 6605055
BC100285.1 - 3, 4 420 6022858
BG829769.1 - 3, 4 261
BM841336.1 - 4, 5 313
D14531.1 - 3, 4 260
U09953.1 - 4, 5 283
U21138.1 - 1 260

Gene Symbol
RPL9P11
Entrez Gene ID
642740
Gene Name
ribosomal protein L9 pseudogene 11
Gene Aliases
RPL9_1_9
Chromosome Location
Chr. 1: 9576405 - 9577081 on Build GRCh38
UniGene ID
Hs.661110
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK130277.1 - 1 889

View Full Product Details