Gene Transcripts

Gene Symbol
Pqlc2
Entrez Gene ID
212555
Gene Name
PQ loop repeat containing 2
Gene Aliases
BC019216
Chromosome Location
Chr. 4: 139294030 - 139310711 on Build GRCm38
UniGene ID
Mm.45131
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145384.2 NP_663359.2 6, 7 803
GenBank mRNA AK081661.1 - 6, 7 790
AK156668.1 - 6, 7 803
BC019216.1 - 6, 7 774 5065163
BC025609.1 - 5, 6 741 5309871

View Full Product Details