Gene Transcripts
- Gene Symbol
- PAM
- Entrez Gene ID
- 5066
- Gene Name
- peptidylglycine alpha-amidating monooxygenase
- Gene Aliases
- PAL, PHM
- Chromosome Location
- Chr. 5: 102754783 - 103031105 on Build GRCh38
- UniGene ID
- Hs.369430
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000919.3 |
NP_000910.2
|
1, 2 |
470 |
|
NM_001177306.1 |
NP_001170777.1
|
1, 2 |
470 |
|
NM_001319943.1 |
NP_001306872.1
|
2, 3 |
1036 |
|
NM_138766.2 |
NP_620121.1
|
1, 2 |
470 |
|
NM_138821.2 |
NP_620176.1
|
1, 2 |
470 |
|
NM_138822.2 |
NP_620177.1
|
1, 2 |
470 |
|
NR_033440.1 |
|
1, 2 |
470 |
|
XM_011543419.2 |
XP_011541721.1
|
2, 3 |
590 |
|
XM_017009495.1 |
XP_016864984.1
|
1, 2 |
466 |
|
XM_017009496.1 |
XP_016864985.1
|
1, 2 |
466 |
|
XM_017009497.1 |
XP_016864986.1
|
2, 3 |
590 |
|
XM_017009498.1 |
XP_016864987.1
|
1, 2 |
466 |
|
XM_017009499.1 |
XP_016864988.1
|
1, 2 |
466 |
|
XM_017009500.1 |
XP_016864989.1
|
1, 2 |
466 |
|
XM_017009501.1 |
XP_016864990.1
|
1, 2 |
466 |
|
XM_017009502.1 |
XP_016864991.1
|
1, 2 |
466 |
|
XM_017009503.1 |
XP_016864992.1
|
1, 2 |
466 |
|
XM_017009504.1 |
XP_016864993.1
|
1, 2 |
567 |
|
XM_017009505.1 |
XP_016864994.1
|
2, 3 |
590 |
|
XM_017009506.1 |
XP_016864995.1
|
1, 2 |
466 |
|
XM_017009507.1 |
XP_016864996.1
|
2, 3 |
590 |
|
XM_017009508.1 |
XP_016864997.1
|
1, 2 |
466 |
|
XM_017009509.1 |
XP_016864998.1
|
1, 2 |
466 |
|
XM_017009510.1 |
XP_016864999.1
|
1, 2 |
570 |
|
XM_017009511.1 |
XP_016865000.1
|
1, 2 |
466 |
|
XM_017009512.1 |
XP_016865001.1
|
1, 2 |
466 |
|
XM_017009513.1 |
XP_016865002.1
|
1, 2 |
466 |
|
XM_017009514.1 |
XP_016865003.1
|
1, 2 |
466 |
|
XM_017009515.1 |
XP_016865004.1
|
1, 2 |
466 |
|
XM_017009516.1 |
XP_016865005.1
|
1, 2 |
466 |
|
GenBank mRNA |
AB095007.1 |
- |
1, 2 |
97 |
|
AF010472.1 |
- |
|
13 |
|
AK290158.1 |
- |
1, 2 |
208 |
|
AK296521.1 |
- |
1, 2 |
236 |
|
BC018127.1 |
- |
1, 2 |
470 |
3887394 |
BT007419.1 |
- |
1, 2 |
97 |
|
DA240822.1 |
- |
2, 3 |
543 |
|
M37721.1 |
- |
1, 2 |
285 |
|
S75037.1 |
- |
1, 2 |
177 |
|
S75038.1 |
- |
1, 2 |
177 |
|
View Full Product Details