Gene Transcripts

Gene Symbol
Plcd3
Entrez Gene ID
72469
Gene Name
phospholipase C, delta 3
Gene Aliases
2610205J15Rik, mKIAA1964
Chromosome Location
Chr. 11: 103069236 - 103101658 on Build GRCm38
UniGene ID
Mm.264743
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152813.3 NP_690026.2 4 759
GenBank mRNA AK046669.1 - 4 725
AK144950.1 - 1832
AK164809.1 - 4 725
AK166977.1 - 4 752
AK173311.1 - 4 731
BC031392.1 - 4 703 4952407

View Full Product Details