Gene Transcripts
- Gene Symbol
- Pygb
- Entrez Gene ID
- 110078
- Gene Name
- brain glycogen phosphorylase
- Gene Aliases
- Chromosome Location
- Chr. 2: 150786796 - 150831748 on Build GRCm38
- UniGene ID
- Mm.222584
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_153781.1 |
NP_722476.1
|
13, 14 |
1691 |
|
GenBank mRNA |
AK032769.1 |
- |
13, 14 |
1704 |
|
AK035673.1 |
- |
13, 14 |
1421 |
|
AK133794.1 |
- |
4, 5 |
421 |
|
AK134667.1 |
- |
13, 14 |
1701 |
|
AK142716.1 |
- |
13, 14 |
1701 |
|
AK147763.1 |
- |
13, 14 |
1723 |
|
AK168381.1 |
- |
13, 14 |
1703 |
|
AK169052.1 |
- |
13, 14 |
1726 |
|
AK169631.1 |
- |
13, 14 |
1705 |
|
BC032209.1 |
- |
7, 8 |
846 |
|
BC035283.1 |
- |
13, 14 |
1691 |
4947455 |
View Full Product Details