Gene Transcripts

Gene Symbol
Hinfp
Entrez Gene ID
102423
Gene Name
histone H4 transcription factor
Gene Aliases
AA589481, HiNF-P, Mizf
Chromosome Location
Chr. 9: 44292438 - 44305705 on Build GRCm38
UniGene ID
Mm.24570
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172162.3 NP_751894.2 10 1998
GenBank mRNA AK049348.1 - 10 1958
BC029091.1 - 9 1981 2651006

View Full Product Details