Gene Transcripts
- Gene Symbol
- Insc
- Entrez Gene ID
- 233752
- Gene Name
- inscuteable homolog (Drosophila)
- Gene Aliases
- 3830422K02Rik, C730020D23
- Chromosome Location
- Chr. 7: 114743677 - 114850380 on Build GRCm38
- UniGene ID
- Mm.83037
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_173767.3 |
NP_776128.2
|
5, 6 |
874 |
|
GenBank mRNA |
AK014452.1 |
- |
4, 5 |
635 |
|
AK050143.1 |
- |
2, 3 |
369 |
|
AK179765.1 |
- |
3, 4 |
257 |
|
AK187129.1 |
- |
1, 2 |
144 |
|
BC026405.1 |
- |
5, 6 |
761 |
4211657 |
BC089586.1 |
- |
2, 3 |
378 |
|
BC139196.1 |
- |
2, 3 |
262 |
8862218 |
BC139197.1 |
- |
2, 3 |
262 |
8862219 |
DQ205645.1 |
- |
4, 5 |
587 |
|
View Full Product Details