Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JAZ5
Entrez Gene ID
838310
Gene Name
jasmonate-zim-domain protein 5
Gene Aliases
AT1G17380, F28G4.16, TIFY11A, jasmonate-zim-domain protein 5
Chromosome Location
Chr.1: 5955155 - 5957511 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332301.1 NP_001320904.1 1 - 2 367 62
NM_101599.3 NP_564019.1 1 - 2 623 62
GenBank mRNA BT000430.1 - 1 - 2 324 62
BT002543.1 - 1 - 2 182 62

View Full Product Details