Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PnsB4
Entrez Gene ID
838455
Gene Name
NDH dependent flow 6
Gene Aliases
AT1G18730, F6A14.16, F6A14_16, NDF6, NDH dependent flow 6, Photosynthetic NDH subcomplex B 4
Chromosome Location
Chr.1: 6460403 - 6462344 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084090.2 NP_001077559.2 3 - 4 313 80
NM_001123834.1 NP_001117306.1 3 - 4 313 80
NM_001123835.4 NP_001117307.1 3 - 4 313 80
NM_001332365.1 NP_001321859.1 1 - 2 233 80
NM_101731.5 NP_173308.2 3 - 4 511 80
GenBank mRNA AK226496.1 - 3 - 4 349 80
AY080701.1 - 3 - 4 348 80
AY117165.1 - 3 - 4 289 80

View Full Product Details