Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXPA11
Entrez Gene ID
838608
Gene Name
expansin 11
Gene Aliases
AT1G20190, ATEXP11, ATEXPA11, ATHEXP ALPHA 1.14, EXP11, EXPANSIN 11, EXPANSIN 11, T20H2.4, T20H2_4, expansin 11
Chromosome Location
Chr.1: 6998335 - 6999839 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332440.1 NP_001320974.1 1 - 2 345 138
NM_101872.4 NP_173446.1 2 - 3 577 138
GenBank mRNA AF332436.1 - 2 - 3 448 138

View Full Product Details