Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARA7
Entrez Gene ID
827706
Gene Name
Ras-related small GTP-binding family protein
Gene Aliases
ARA-7, ARABIDOPSIS RAB GTPASE HOMOLOG F2B, AT4G19640, ATRAB-F2B, ATRAB5B, ATRABF2B, F24J7.190, F24J7_190, RAB GTPASE HOMOLOG F2B, RAB-F2B, RABF2B
Chromosome Location
Chr.4: 10687196 - 10689748 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118084.5 NP_193699.1 5 - 6 709 114
GenBank mRNA AB038491.1 - 5 - 6 410 114
AF370309.1 - 5 - 6 536 114
AY052670.1 - 5 - 6 572 114
AY063095.1 - 5 - 6 410 114

View Full Product Details